Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317739.1 | NP_001304668.1 | 2 | 262 | ||
NM_005868.5 | NP_005859.1 | 2 | 262 | |||
NR_133908.1 | 2 | 262 | ||||
NR_133909.1 | 2 | 262 | ||||
GenBank mRNA | AF007551.1 | - | 2 | 215 | ||
AK313984.1 | - | 2 | 235 | |||
BC000899.1 | - | 2 | 151 | 2899921 | ||
BC012595.1 | - | 2 | 245 | 4096064 | ||
BG721261.1 | - | 2 | 262 | |||
BT009853.1 | - | 2 | 96 | |||
BX505568.1 | - | 2 | 262 | |||
CR749267.1 | - | 2 | 213 | |||
DA127369.1 | - | 2 | 235 |