Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRND | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012409.3 | NP_036541.2 | 2 | 225 | ||
GenBank mRNA | AF187843.1 | - | 2 | 159 | ||
AF187844.1 | - | 1 | 110 | |||
AK313404.1 | - | 2 | 226 | |||
AY358985.1 | - | 2 | 154 | |||
BC043644.2 | - | 2 | 169 | 5272576 |