Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014047.2 | NP_054766.1 | 1 | 107 | ||
GenBank mRNA | AF078852.1 | - | 1 | 81 | ||
AF086483.1 | - | 1 | 79 | |||
AK311855.1 | - | 1 | 72 | |||
BC015465.2 | - | 1 | 82 | 3914049 | ||
DA768829.1 | - | 1 | 108 |