Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CRISP2 SIRNA Assays ›
Gene Symbol:
CRISP2
Gene Name:
cysteine rich secretory protein 2
Gene Aliases:
CRISP-2, CT36, GAPDL5, TPX1, TSP1
Chromosome Location:
Chr.6: 49676665 - 49714198 on Build GRCh38
UniGene ID:
Hs.2042
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4629
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CRISP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142407.2
NP_001135879.1
5
409
NM_001142408.2
NP_001135880.1
6
466
NM_001142417.2
NP_001135889.1
6
446
NM_001142435.2
NP_001135907.1
5
429
NM_001261822.1
NP_001248751.1
5
435
NM_003296.3
NP_003287.1
6
472
XM_005249349.1
XP_005249406.1
6
446
XM_005249350.1
XP_005249407.1
5
404
XM_005249351.3
XP_005249408.1
5
424
XM_005249352.3
XP_005249409.1
5
415
XM_005249353.3
XP_005249410.1
6
451
XM_005249356.1
XP_005249413.1
6
449
XM_011514841.1
XP_011513143.1
6
781
XM_011514842.1
XP_011513144.1
5
744
XM_011514843.2
XP_011513145.1
6
451
XM_011514844.1
XP_011513146.1
6
451
GenBank mRNA
AK223337.1
-
6
409
AK292385.1
-
6
450
AY289796.1
-
6
416
AY292862.1
-
5
406
AY292863.1
-
5
410
AY773479.1
-
6
452
BC022011.2
-
6
419
4826427
BC107707.1
-
5
366
6618679
BG718726.1
-
5
405
BI562149.1
-
5
392
BM552843.1
-
5
344
BP369378.1
-
5
422
DB082240.1
-
6
446
DB459032.1
-
6
442
M25532.1
-
6
395
X95239.1
-
6
446
Back To Top
More Information
Panther Classification:
Molecular Function -
defense/immunity protein
Gene Ontology Categories:
Function(s)
single organismal cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline