Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVRIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024070.3 | NP_076975.2 | 6 | 1425 | ||
XM_011516575.2 | XP_011514877.1 | 7 | 1441 | |||
GenBank mRNA | BC001129.2 | - | 5 | 1117 | ||
BC032113.2 | - | 3 | 816 | |||
BC073861.1 | - | 5 | 1088 | |||
BC107873.1 | - | 7 | 1450 | 6452536 |