Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MRPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031903.2 | NP_114109.1 | 2, 3 | 362 | ||
GenBank mRNA | AB049649.1 | - | 2, 3 | 361 | ||
AF161401.1 | - | 2, 3 | 302 | |||
AK313393.1 | - | 2, 3 | 320 | |||
BC013147.1 | - | 2, 3 | 321 | 4340082 | ||
BM833110.1 | - | 2, 3 | 325 |