Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM184A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097620.1 | NP_001091089.1 | 3 | 662 | ||
XM_006715653.1 | XP_006715716.1 | 3 | 662 | |||
XM_006715655.2 | XP_006715718.1 | 3 | 505 | |||
XM_011515164.1 | XP_011513466.1 | 3 | 662 | |||
XM_011515165.2 | XP_011513467.1 | 3 | 662 | |||
XM_011515166.2 | XP_011513468.1 | 3 | 662 | |||
XM_017011815.1 | XP_016867304.1 | 3 | 505 | |||
GenBank mRNA | AK075043.1 | - | 3 | 520 | ||
AK096790.1 | - | 1 | 74 | |||
AK126312.1 | - | 3 | 535 | |||
AK172850.1 | - | 3 | 438 | |||
AK313249.1 | - | 3 | 446 | |||
BC026694.1 | - | 3 | 437 | 3850351 |