Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ninj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012867.1 | NP_036999.1 | 3 | 319 | ||
GenBank mRNA | FQ215609.1 | - | 3 | 351 | ||
FQ219073.1 | - | 3 | 351 | |||
FQ220812.1 | - | 3 | 351 | |||
FQ225610.1 | - | 3 | 323 | |||
FQ229388.1 | - | 3 | 351 | |||
FQ230574.1 | - | 3 | 323 | |||
FQ232057.1 | - | 3 | 351 | |||
U72660.1 | - | 3 | 341 |