Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019207.1 | NP_062080.1 | 1 | 563 | ||
GenBank mRNA | U67777.1 | - | 1 | 563 |