Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012812.3 | NP_036944.2 | 3 | 374 | ||
XM_006230218.1 | XP_006230280.1 | 4 | 423 | |||
GenBank mRNA | FM058713.1 | - | 4 | 428 | ||
FM121690.1 | - | 3 | 280 | |||
FQ217951.1 | - | 3 | 324 | |||
FQ217987.1 | - | 3 | 281 | |||
FQ223608.1 | - | 3 | 330 | |||
FQ224015.1 | - | 3 | 332 | |||
FQ224196.1 | - | 3 | 331 | |||
FQ224467.1 | - | 3 | 320 | |||
FQ224572.1 | - | 3 | 339 | |||
X12554.1 | - | 3 | 295 |