Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: App | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019288.2 | NP_062161.1 | 2 | 204 | ||
XM_006248008.2 | XP_006248070.1 | 2 | 334 | |||
XM_006248009.2 | XP_006248071.1 | 2 | 334 | |||
XM_006248010.2 | XP_006248072.1 | 2 | 334 | |||
XM_006248011.2 | XP_006248073.1 | 2 | 334 | |||
XM_006248012.2 | XP_006248074.1 | 2 | 334 | |||
GenBank mRNA | AF513015.1 | - | 2 | 205 | ||
BC062082.1 | - | 2 | 317 | 5599880 | ||
FQ221425.1 | - | 2 | 337 |