Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nppc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053750.1 | NP_446202.1 | 2 | 260 | ||
GenBank mRNA | D90219.1 | - | 2 | 260 |