Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF4 SIRNA Assays ›
Gene Symbol:
TCF4
Gene Name:
transcription factor 4
Gene Aliases:
E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
Chromosome Location:
Chr.18: 55222331 - 55635993 on Build GRCh38
UniGene ID:
Hs.605153
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4699
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083962.1
NP_001077431.1
17
2210
NM_001243226.2
NP_001230155.2
18
2000
NM_001243227.1
NP_001230156.1
16
2044
NM_001243228.1
NP_001230157.1
17
2154
NM_001243230.1
NP_001230159.1
16
1683
NM_001243231.1
NP_001230160.1
15
1644
NM_001243232.1
NP_001230161.1
13
1666
NM_001243233.1
NP_001230162.1
13
1607
NM_001243234.1
NP_001230163.1
10
1239
NM_001243235.1
NP_001230164.1
10
1239
NM_001243236.1
NP_001230165.1
10
1255
NM_001306207.1
NP_001293136.1
16
1835
NM_001306208.1
NP_001293137.1
13
1666
NM_003199.2
NP_003190.1
17
2210
XM_005266739.3
XP_005266796.2
16
1850
XM_005266741.3
XP_005266798.1
17
1810
XM_005266743.4
XP_005266800.1
16
1671
XM_005266744.3
XP_005266801.1
16
1791
XM_005266745.3
XP_005266802.1
16
1653
XM_005266747.3
XP_005266804.1
15
1647
XM_005266749.3
XP_005266806.1
13
1438
XM_005266752.4
XP_005266809.1
13
2008
XM_005266754.3
XP_005266811.1
12
1432
XM_005266755.4
XP_005266812.1
13
1467
XM_005266761.3
XP_005266818.1
10
1748
XM_006722536.2
XP_006722599.1
17
2133
XM_006722537.2
XP_006722600.1
17
1766
XM_006722538.2
XP_006722601.1
16
1788
XM_011526158.2
XP_011524460.1
16
2764
XM_011526160.2
XP_011524462.1
12
1817
XM_017025934.1
XP_016881423.1
16
1847
XM_017025935.1
XP_016881424.1
16
1847
XM_017025936.1
XP_016881425.1
16
1850
XM_017025937.1
XP_016881426.1
16
1604
XM_017025938.1
XP_016881427.1
17
1763
XM_017025939.1
XP_016881428.1
17
1810
XM_017025940.1
XP_016881429.1
17
2133
XM_017025941.1
XP_016881430.1
17
1766
XM_017025942.1
XP_016881431.1
17
1810
XM_017025943.1
XP_016881432.1
17
1810
XM_017025944.1
XP_016881433.1
16
2068
XM_017025945.1
XP_016881434.1
16
2068
XM_017025946.1
XP_016881435.1
16
2085
XM_017025947.1
XP_016881436.1
16
1671
XM_017025948.1
XP_016881437.1
16
2068
XM_017025949.1
XP_016881438.1
16
1788
XM_017025950.1
XP_016881439.1
16
2065
XM_017025951.1
XP_016881440.1
15
1644
XM_017025952.1
XP_016881441.1
13
1658
XM_017025953.1
XP_016881442.1
13
1438
XM_017025954.1
XP_016881443.1
13
2019
XM_017025955.1
XP_016881444.1
12
1753
XM_017025956.1
XP_016881445.1
12
1432
XM_017025957.1
XP_016881446.1
10
1748
GenBank mRNA
AB209741.1
-
12
1318
AK095041.1
-
13
1666
AK096862.1
-
15
1644
AK122765.1
-
17
2210
AK299169.1
-
16
2044
AK300038.1
-
10
1185
AK300612.1
-
13
1607
AK300636.1
-
12
1337
AK301144.1
-
16
1821
AK315074.1
-
18
2142
AK316165.1
-
12
1381
BC031056.1
-
18
1961
BC125084.1
-
16
1624
40117795
BC125085.1
-
16
1624
40117798
FR748210.1
-
16
1598
FR748211.1
-
16
1598
FR748212.1
-
14
1418
FR748213.1
-
14
1418
FR748214.1
-
15
1526
FR748215.1
-
13
1346
FR748216.1
-
16
1589
FR748217.1
-
15
1472
FR748218.1
-
11
1208
FR748219.1
-
13
1385
FR748220.1
-
10
1118
FR748221.1
-
10
1118
FR748222.1
-
10
1118
FR748223.1
-
8
950
M74718.1
-
10
1070
M74719.1
-
17
1797
X52079.1
-
14
1463
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001083967, NM_013685, U16321, AK028748, AK081012, and others.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein-DNA complex assembly
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, TFIIB-class binding
TFIIB-class transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein C-terminus binding
identical protein binding
protein homodimerization activity
bHLH transcription factor binding
protein heterodimerization activity
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0