Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053983.1 | NP_446435.1 | 2 | 370 | ||
GenBank mRNA | X76697.1 | - | 2 | 370 |