Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057200.2 | NP_476548.1 | 2, 3 | 469 | ||
XM_008764204.1 | XP_008762426.1 | 2, 3 | 475 | |||
GenBank mRNA | AB070355.1 | - | 2, 3 | 469 | ||
AB120429.1 | - | 1, 2 | 115 | |||
AF083331.1 | - | 1, 2 | 115 |