Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC257642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147136.1 | NP_671477.1 | 616 | |||
GenBank mRNA | U77931.1 | - | 616 |
Gene Symbol: LOC103693757 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770183.1 | XP_008768405.1 | 4 | 2136 |