Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142407.2 | NP_001135879.1 | 9 | 1096 | ||
NM_001142408.2 | NP_001135880.1 | 10 | 1153 | |||
NM_001142417.2 | NP_001135889.1 | 10 | 1133 | |||
NM_001142435.2 | NP_001135907.1 | 9 | 1116 | |||
NM_001261822.1 | NP_001248751.1 | 9 | 1122 | |||
NM_003296.3 | NP_003287.1 | 10 | 1159 | |||
XM_005249349.1 | XP_005249406.1 | 11 | 1238 | |||
XM_005249350.1 | XP_005249407.1 | 10 | 1196 | |||
XM_005249351.3 | XP_005249408.1 | 10 | 1216 | |||
XM_005249352.3 | XP_005249409.1 | 10 | 1207 | |||
XM_005249353.3 | XP_005249410.1 | 11 | 1243 | |||
XM_005249356.1 | XP_005249413.1 | 11 | 1241 | |||
XM_011514841.1 | XP_011513143.1 | 11 | 1573 | |||
XM_011514842.1 | XP_011513144.1 | 10 | 1536 | |||
GenBank mRNA | AK128009.1 | - | 1 | 2089 | ||
AK223337.1 | - | 10 | 1096 | |||
AK292385.1 | - | 10 | 1137 | |||
AY289796.1 | - | 10 | 1103 | |||
AY292862.1 | - | 10 | 1198 | |||
AY292863.1 | - | 9 | 1097 | |||
AY773479.1 | - | 10 | 1139 | |||
BC022011.2 | - | 10 | 1106 | 4826427 | ||
BC107707.1 | - | 9 | 1053 | 6618679 | ||
BI831679.1 | - | 4 | 508 | |||
DB524518.1 | - | 1 | 225 | |||
M25532.1 | - | 10 | 1082 | |||
X95239.1 | - | 10 | 1133 |