Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173133.2 | NP_775156.2 | 2 | 210 | ||
GenBank mRNA | BC086410.1 | - | 2 | 180 | ||
BC097932.1 | - | 2 | 186 | 7386998 | ||
X67877.1 | - | 2 | 210 |