Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171990.2 | NP_741987.1 | 5, 6 | 466 | ||
XM_006250429.2 | XP_006250491.1 | 5, 6 | 508 | |||
XM_006250430.2 | XP_006250492.1 | 5, 6 | 563 | |||
XM_006250431.2 | XP_006250493.1 | 5, 6 | 562 | |||
GenBank mRNA | AJ000347.1 | - | 5, 6 | 417 | ||
BC085692.1 | - | 5, 6 | 466 | 7190529 | ||
FQ214355.1 | - | 5, 6 | 489 |