Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxmp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172223.2 | NP_757377.1 | 3, 4 | 424 | ||
GenBank mRNA | AY151834.1 | - | 3, 4 | 402 | ||
BC081699.1 | - | 3, 4 | 424 | 7193564 |