Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271058.1 | NP_001257987.1 | 5 | 611 | ||
NM_001271059.1 | NP_001257988.1 | 6 | 655 | |||
NM_001271060.1 | NP_001257989.1 | 5 | 646 | |||
NM_001271061.1 | NP_001257990.1 | 5 | 580 | |||
NM_001271062.1 | NP_001257991.1 | 5 | 689 | |||
NM_173096.3 | NP_775119.2 | 5 | 769 | |||
XM_006248146.2 | XP_006248208.1 | 6 | 835 | |||
XM_006248149.2 | XP_006248211.1 | 6 | 878 | |||
XM_008768555.1 | XP_008766777.1 | 5 | 2771 | |||
XM_008768556.1 | XP_008766778.1 | 5 | 2744 | |||
XM_008768557.1 | XP_008766779.1 | 5 | 2679 | |||
GenBank mRNA | BC099784.1 | - | 5 | 757 | 7933587 | |
FQ226931.1 | - | 5 | 646 | |||
FQ230542.1 | - | 5 | 658 | |||
FQ231581.1 | - | 6 | 723 | |||
X52711.1 | - | 4 | 563 |