Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175579.3 | NP_783169.2 | 1 | 227 | ||
XM_006247278.2 | XP_006247340.1 | 1 | 332 | |||
XM_006247279.2 | XP_006247341.1 | 1 | 332 | |||
GenBank mRNA | AY187039.1 | - | 1 | 204 | ||
AY192567.1 | - | 1 | 227 |