Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175763.2 | NP_786939.1 | 4 | 267 | ||
XM_006257557.2 | XP_006257619.1 | 5 | 361 | |||
XM_006257558.1 | XP_006257620.1 | 4 | 311 | |||
XM_006257559.1 | XP_006257621.1 | 4 | 314 | |||
XM_006257560.1 | XP_006257622.1 | 4 | 314 | |||
XM_006257561.1 | XP_006257623.1 | 4 | 314 | |||
XM_008773563.1 | XP_008771785.1 | 4 | 314 | |||
GenBank mRNA | AF322216.1 | - | 4 | 267 | ||
AF322217.1 | - | 4 | 265 | |||
BC060309.1 | - | 4 | 294 | |||
BC127478.1 | - | 4 | 241 |