Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177482.1 | NP_803435.1 | 2 | 208 | ||
XM_008760227.1 | XP_008758449.1 | 4 | 532 | |||
XM_008760228.1 | XP_008758450.1 | 4 | 663 | |||
GenBank mRNA | BC099696.1 | - | 2 | 188 | ||
J00774.1 | - | 1 | 128 | |||
V01255.1 | - | 2 | 225 | |||
V01545.1 | - | 2 | 208 |