Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175757.2 | NP_786933.1 | 2 | 164 | ||
GenBank mRNA | AY040223.1 | - | 2 | 164 | ||
BC078685.1 | - | 2 | 138 | 7110482 |