Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012530.2 | NP_036662.1 | 3 | 341 | ||
GenBank mRNA | DN931989.1 | - | 3 | 340 | ||
FQ214594.1 | - | 3 | 317 | |||
FQ214655.1 | - | 3 | 317 | |||
FQ214750.1 | - | 3 | 317 | |||
FQ214825.1 | - | 2 | 138 | |||
FQ214931.1 | - | 3 | 318 | |||
FQ215369.1 | - | 2 | 215 | |||
FQ215410.1 | - | 3 | 317 | |||
FQ215927.1 | - | 2 | 170 | |||
FQ216321.1 | - | 2 | 169 | |||
FQ216507.1 | - | 3 | 312 | |||
FQ217421.1 | - | 2 | 182 | |||
FQ224848.1 | - | 2 | 168 | |||
FQ224918.1 | - | 3 | 318 | |||
M10140.1 | - | 3 | 314 |