Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iapp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012586.2 | NP_036718.1 | 2 | 251 | ||
XM_006237614.2 | XP_006237676.1 | 3 | 780 | |||
GenBank mRNA | J04544.1 | - | 3 | 338 | ||
M25390.1 | - | 3 | 287 |