Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253599.2 | XP_006253661.1 | 2 | 1099 | ||
XM_006253600.2 | XP_006253662.1 | 2 | 912 | |||
GenBank mRNA | M35077.1 | - | 1 | 235 |
Set Membership: |
G-protein coupled receptor |