Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012575.3 | NP_036707.3 | 4, 5 | 1407 | ||
XM_006247706.2 | XP_006247768.1 | 2, 3 | 1126 | |||
XM_006247707.1 | XP_006247769.1 | 5, 6 | 1568 | |||
XM_006247708.2 | XP_006247770.1 | 2, 3 | 1023 | |||
XM_006247709.2 | XP_006247771.1 | 2, 3 | 1023 | |||
GenBank mRNA | D13212.1 | - | 4, 5 | 1407 | ||
M91563.1 | - | 2, 3 | 1026 | |||
U08259.1 | - | 5, 6 | 1567 |