Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Andpro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012718.2 | NP_036850.1 | 1 | 149 | ||
XM_006235184.2 | XP_006235246.1 | 1 | 388 | |||
GenBank mRNA | BC065311.1 | - | 1 | 129 | ||
M27901.1 | - | 1 | 133 | |||
M58167.1 | - | 1 | 150 |