Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012769.2 | NP_036901.2 | 3 | 199 | ||
GenBank mRNA | AB098025.1 | - | 2 | 111 | ||
AB098707.1 | - | 2 | 111 | |||
AB099520.1 | - | 2 | 111 | |||
AB099521.2 | - | 2 | 111 | |||
BC081840.1 | - | 3 | 199 | 7104600 | ||
M22562.1 | - | 3 | 152 |