Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012778.1 | NP_036910.1 | 1 | 209 | ||
GenBank mRNA | BC090068.1 | - | 1 | 191 | 7190406 | |
L07268.1 | - | 1 | 200 | |||
X67948.1 | - | 1 | 209 | |||
X70257.1 | - | 1 | 149 | |||
X71069.1 | - | 1 | 160 |