Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012733.4 | NP_036865.1 | 2 | 318 | ||
GenBank mRNA | FQ209770.1 | - | 2 | 332 | ||
FQ209927.1 | - | 2 | 302 | |||
FQ210835.1 | - | 2 | 333 | |||
FQ218090.1 | - | 2 | 332 | |||
M16459.1 | - | 2 | 318 | |||
M19257.1 | - | 2 | 248 |