Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012747.2 | NP_036879.1 | 4 | 492 | ||
XM_006247257.2 | XP_006247319.1 | 4 | 516 | |||
XM_006247258.2 | XP_006247320.1 | 4 | 516 | |||
XM_006247259.2 | XP_006247321.1 | 4 | 516 | |||
GenBank mRNA | BC087025.1 | - | 4 | 492 | 7110655 | |
X91810.1 | - | 3 | 286 |