Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012810.1 | NP_036942.1 | 6 | 455 | ||
GenBank mRNA | X67805.1 | - | 6 | 455 |