Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100363178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749374.3 | XP_003749422.1 | 964 |
Gene Symbol: Adora3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF102804.1 | - | 1 | 142 | ||
DQ075463.1 | - | 1 | 145 | |||
M94152.1 | - | 460 | ||||
X59249.1 | - | 1 | 273 | |||
X93219.1 | - | 1 | 145 |
Set Membership: |
G-protein coupled receptor |