Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257198.1 | NP_001244127.1 | 2, 3 | 343 | ||
NM_003590.4 | NP_003581.1 | 2, 3 | 660 | |||
XM_006712800.2 | XP_006712863.2 | 2, 3 | 333 | |||
XM_011511995.1 | XP_011510297.1 | 2, 3 | 245 | |||
XM_011511996.2 | XP_011510298.1 | 2, 3 | 309 | |||
GenBank mRNA | AB014517.2 | - | 2, 3 | 588 | ||
AF062537.1 | - | 2, 3 | 361 | |||
AF064087.1 | - | 2, 3 | 353 | |||
AK291151.1 | - | 2, 3 | 310 | |||
AK308496.1 | - | 2, 3 | 343 | |||
AY337761.1 | - | 2, 3 | 806 | |||
BC039598.1 | - | 2, 3 | 605 | 5784147 | ||
BC092409.1 | - | 2, 3 | 605 | 6168545 | ||
BQ688156.1 | - | 660 |