Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012903.1 | NP_037035.1 | 6 | 761 | ||
XM_008766210.1 | XP_008764432.1 | 6 | 905 | |||
XM_008766211.1 | XP_008764433.1 | 6 | 695 | |||
XM_008766212.1 | XP_008764434.1 | 6 | 811 | |||
XM_008766213.1 | XP_008764435.1 | 6 | 637 | |||
GenBank mRNA | BC087686.1 | - | 6 | 764 | 7308609 | |
D32209.1 | - | 6 | 761 |
Gene Symbol: LOC100909983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773869.1 | XP_008772091.1 | 614 |