Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012924.2 | NP_037056.2 | 2 | 223 | ||
XM_006234627.2 | XP_006234689.1 | 2 | 368 | |||
XM_006234629.2 | XP_006234691.1 | 2 | 365 | |||
XM_006234631.2 | XP_006234693.1 | 2 | 342 | |||
XM_008762055.1 | XP_008760277.1 | 2 | 367 | |||
XM_008762056.1 | XP_008760278.1 | 2 | 365 | |||
XM_008762057.1 | XP_008760279.1 | 2 | 363 | |||
XM_008762058.1 | XP_008760280.1 | 2 | 357 | |||
XM_008762059.1 | XP_008760281.1 | 2 | 372 | |||
GenBank mRNA | AF065147.1 | - | 2 | 135 | ||
BC061531.1 | - | 2 | 227 | |||
BC085703.1 | - | 2 | 253 | |||
BC107949.1 | - | 2 | 208 | |||
BC127485.1 | - | 2 | 206 | |||
FQ234376.1 | - | 2 | 223 | |||
M61874.1 | - | 2 | 223 | |||
M61875.1 | - | 2 | 223 | |||
U46957.1 | - | 2 | 135 | |||
U52179.1 | - | 2 | 134 | |||
U96138.1 | - | 2 | 223 |