Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pou1f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013008.3 | NP_037140.2 | 2 | 272 | ||
XM_006247974.1 | XP_006248036.1 | 2 | 556 | |||
GenBank mRNA | AB622208.1 | - | 1 | 283 | ||
BC061563.1 | - | 2 | 276 | 6918965 | ||
BC061564.1 | - | 2 | 291 | |||
L01506.1 | - | 2 | 240 | |||
L01507.1 | - | 2 | 316 | |||
M23253.1 | - | 2 | 186 | |||
X12658.1 | - | 2 | 209 |