Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012922.2 | NP_037054.1 | 5 | 379 | ||
XM_006253130.2 | XP_006253192.1 | 5 | 423 | |||
GenBank mRNA | BC081854.1 | - | 5 | 379 | 7104685 | |
U34685.1 | - | 3 | 237 | |||
U49930.1 | - | 4 | 349 | |||
U84410.1 | - | 4 | 353 |