Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012974.1 | NP_037106.1 | 6, 7 | 728 | ||
XM_006243709.2 | XP_006243771.1 | 6, 7 | 773 | |||
GenBank mRNA | BC088400.1 | - | 6, 7 | 727 | 7107264 | |
X16563.1 | - | 6, 7 | 728 |