Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277226.1 | NP_001264155.1 | 5 | 859 | ||
NM_003667.3 | NP_003658.1 | 5 | 859 | |||
NR_110596.1 | 5 | 859 | ||||
GenBank mRNA | AB105190.1 | - | 5 | 828 | ||
AF061444.1 | - | 5 | 548 | |||
AF062006.1 | - | 5 | 596 | |||
AK075399.1 | - | 5 | 645 | |||
AK294985.1 | - | 5 | 860 | |||
AK315133.1 | - | 5 | 836 | |||
BC096324.3 | - | 5 | 594 | 40008253 | ||
BC096325.1 | - | 5 | 594 | 40008255 | ||
BC096326.3 | - | 5 | 594 | 40008257 | ||
BC099650.1 | - | 5 | 594 | 40008256 |