Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012988.2 | NP_037120.1 | 1 | 334 | ||
GenBank mRNA | AB060652.1 | - | 242 | |||
AF112455.1 | - | 414 | ||||
AF112456.1 | - | 1 | 127 | |||
AY572794.1 | - | 1 | 242 | |||
D78017.1 | - | 1 | 334 | |||
D78018.1 | - | 1 | 329 | |||
D78019.1 | - | 1 | 314 | |||
X13167.1 | - | 1 | 176 |