Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012993.2 | NP_037125.1 | 2 | 431 | ||
XM_006238499.2 | XP_006238561.1 | 2 | 565 | |||
XM_006238500.2 | XP_006238562.1 | 2 | 563 | |||
GenBank mRNA | FQ218095.1 | - | 2 | 554 | ||
L27124.1 | - | 2 | 432 | |||
X93208.1 | - | 2 | 432 |