Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asns | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013079.2 | NP_037211.2 | 2 | 381 | ||
XM_006236089.2 | XP_006236151.1 | 2 | 374 | |||
GenBank mRNA | BC081719.1 | - | 2 | 381 | 7132312 | |
U07201.1 | - | 2 | 325 | |||
U07202.1 | - | 2 | 325 |