Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003842.4 | NP_003833.4 | 3, 4 | 654 | ||
NM_147187.2 | NP_671716.2 | 3, 4 | 654 | |||
NR_027140.1 | 2, 3 | 548 | ||||
XM_017013944.1 | XP_016869433.1 | 3, 4 | 442 | |||
GenBank mRNA | AB451272.1 | - | 3, 4 | 361 | ||
AB451399.1 | - | 3, 4 | 361 | |||
AF012535.1 | - | 3, 4 | 500 | |||
AF012628.1 | - | 3, 4 | 361 | |||
AF016266.1 | - | 3, 4 | 478 | |||
AF016268.1 | - | 3, 4 | 470 | |||
AF016849.1 | - | 3, 4 | 505 | |||
AF018657.1 | - | 3, 4 | 361 | |||
AF018658.1 | - | 3, 4 | 361 | |||
AF020501.1 | - | 3, 4 | 361 | |||
AF022386.1 | - | 3, 4 | 361 | |||
AF153687.1 | - | 3, 4 | 479 | |||
AF192548.1 | - | 3, 4 | 461 | |||
AK291216.1 | - | 3, 4 | 481 | |||
AK298602.1 | - | 2, 3 | 392 | |||
AL529460.3 | - | 3, 4 | 654 | |||
AY358277.1 | - | 3, 4 | 500 | |||
BC001281.1 | - | 3, 4 | 508 | 3458466 | ||
CR541898.1 | - | 3, 4 | 361 | |||
DA738472.1 | - | 3, 4 | 654 | |||
DA889488.1 | - | 3, 4 | 498 | |||
FJ200482.1 | - | 2, 3 | 255 |