Gene Symbol: Ppp1cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013065.2 | NP_037197.1 | 3 | 507 | ||
XM_008764505.1 | XP_008762727.1 | 4 | 445 | |||
GenBank mRNA | BC062033.1 | - | 3 | 507 | 5623630 | |
D90164.1 | - | 3 | 287 | |||
FQ212294.1 | - | 4 | 445 | |||
S78218.1 | - | 3 | 274 |