Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013081.2 | NP_037213.1 | 3, 4 | 794 | ||
XM_008765546.1 | XP_008763768.1 | 5, 6 | 923 | |||
GenBank mRNA | AF020777.1 | - | 3, 4 | 793 |