Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013083.2 | NP_037215.1 | 3, 4 | 543 | ||
GenBank mRNA | BC062017.1 | - | 3, 4 | 503 | 5621611 | |
CB578015.1 | - | 3, 4 | 543 | |||
M14050.1 | - | 2, 3 | 419 |